missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 4, Red, Reagecon™
BIOPHARMA

Catalog Number p-8014505
Click to view available options
Quantity:
250 mL
500 mL
1 L
5 L
Packaging:
Bag-in-Box Container
HDPE Bottle GL45
HDPE Twin Neck Bottle
pH:
4
This item is not returnable. View return policy
This item is not returnable. View return policy

Colored pH Buffer solutions in Twin-neck containers. Reagecon™ Buffer Solution pH 4, Red  are ISO 17025 accredited and NIST traceable.

  • Colored
  • Twin-neck containers with integrated calibrating chamber
  • Tested at 20°C or 25°C
  • Certified by ISO 17025 Accredited Test Method
  • NIST traceable
  • Presented in various pack sizes
Color Red
pH 4
Quantity 250 mL
Packaging HDPE Twin Neck Bottle
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'