missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH9.2 (Borate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™

Brand:  Fisher Chemical J/2870/21

Code : 23

Additional Details : Weight : 5.00000kg

Packaging Plastic container
Quantity 5L

 View more versions of this product

Product Code. 10030180

  • 112.00 € / 5 L

Estimated Shipment
04-03-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

163147

Specifications

163147
Plastic container
Colorless
Liquid
9.2
5L
Videos
SDS Documents