Learn More
Thermo Scientific™ Dry-blend Buffered Packs
Our quality control ensures that every pack of dry-blend buffers yields consistent solutions without weighing or pH adjustment. Simply empty contents of one foil envelope pack into a beaker, add appropriate solvents, and stir to dissolve.
Brand: Thermo Scientific™ 28390
123.28 EUR valid until 2024-03-30
Use promo code "21648" to get your promotional price.
Code : A0
Additional Details : Weight : 0.10000kg
Description
Our dry-blend buffer packs are easy to use. Simply empty contents of one foil envelope pack into a beaker, add ultrapure water, and stir to dissolve. No need to make and store large volumes of stock solution in advance of daily needs.
Benefits of ready-to-use buffered packs include:• Convenient—no weighing, no pH adjustment, no need to stock individual components
• Consistent quality—our quality control ensures that every pack yields the same, consistent buffer
• Long shelf life—stocking and storage as dry packs eliminates concerns about long-term stability of stock solutions
BupH™ Phosphate-Buffered Saline Packs are pouches of dry-blend powder that make sodium phosphate buffered saline (PBS) for use in crosslinking and labeling reactions requiring amine-free conditions. Each BupH Dry-Blend pack makes ready-to-use PBS with 0.1 M sodium phosphate, 0.15 M NaCl buffer at pH 7.2 when dissolved in 500 mL of water.
BupH MES-Buffered Saline Packs are pouches of dry-blended powder that make optimized amine-free and carboxyl-free buffer. When dissolved in 500 mL of water, the buffer contains 0.1 M 2-(N-morpholino) ethanesulfonic acid (MES), 0.9% sodium chloride at pH 4.7 for carbodiimide (EDC) crosslinking and conjugation reactions.
BupH Carbonate-Bicarbonate Buffer Packs are pouches of dry-blended powder that are each sufficient to make 500 mL of alkaline (pH 9.4) sodium-carbonate (carb-bicarb) buffer. This buffer is used for coating or binding capture-antibodies and other proteins onto polystyrene microplates for ELISA and other plate assays. When dissolved in 500 mL of water, each pack makes 0.2 M carbonate-bicarbonate at pH 9.4.
BupH Tris-Buffered Saline Packs are pouches of dry-blend powder that make Tris-NaCl buffered saline (TBS) for diluents and wash buffers in ELISA, western blotting, and general immunoassay procedures. Each pack makes ready-to-use TBS with 25 mM Tris, 0.15 M NaCl at pH 7.2 when dissolved in 500 mL of water.
BupH Borate Buffer Packs are pouches of dry-blended powder that are each sufficient to make borate buffer for use in protein modification procedures requiring amine-free conditions. When dissolved in 500 mL of water, each pack makes 50 mM borate at pH 8.5.
BupH Tris-Glycine Transfer Buffer Packs are pouches of dry-blend powder that make 500 mL of standard transfer buffer for wet or semi-dry electrophoretic protein transfer from gel to blotting membrane. Each pack makes 25 mM Tris, 192 mM glycine at pH 8 when dissolved in 400 mL of water and 100 mL methanol.
BupH Modified Dulbecco's PBS Packs are pouches of dry-blend powder that make physiological sodium- and potassium-phosphate buffered saline (D-PBS) for ELISA and western blot diluents and wash buffers. Each pack makes 8 mM sodium phosphate, 2 mM potassium phosphate, 0.14 M NaCl, 10 mM KCl modified Dulbecco's PBS buffer at pH 7.4 when dissolved in 500 mL of water.
Specifications
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20–25°C. | |
BupH™ | |
10 Packs | |
4.7 | |
Powder |
Dry Blend | |
MES Buffered Saline | |
Carboxyl-to-Amine Crosslinking | |
MES Buffer |
Safety and Handling
- MES Buffered Saline Pack
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 4-Morpholineethanesulfonic acid hydrate,sodium chloride