missing translation for 'onlineSavingsMsg'
Learn More

Butvar(R) B-98, Thermo Scientific Chemicals

Catalog Number p-4090010
Click to view available options
Quantity:
250 g
500 g
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
63148-65-2
MFCD00084458
Poly(vinyl butyral) resin
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
63148-65-2
MFCD00084458
Poly(vinyl butyral) resin

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Density 1.1000g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 250 g
Specific Gravity 1.1
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform
Viscosity 6 to 9cPs (6% in methanol, 20°C)
Percent Purity 18.0 to 20.0% (Hydroxyl content) as PV alcohol, 2.5% max. (Acetate content) as PV acetate
Physical Form Free-flowing Powder
Chemical Name or Material Butvar* B-98
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'