Learn More
Jaytec Glass™ Butyrometers Tube
Brand:  Jaytec Glass™ MP891
Code : 03
Additional Details : Weight : 0.01000kg
Product Code. 12398955
  • 52.05 € / Each
Estimated Shipment: 05-07-2024
to see stock.
Add to basket
Description
Description
Manufactured to BS 696.
For testing fat content in dairy products, using the Gerber method.
All products are for research use only unless otherwise indicated.
Specifications
Specifications
5 g