missing translation for 'onlineSavingsMsg'
Learn More

Butyryl chloride, 98%

Product Code. 11418207
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 11418207

Brand: Thermo Scientific Alfa Aesar A17575.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 141-75-3 | C4H7ClO | 106.55 g/mol

Butyryl chloride is used as an intermediate for the synthesis of active pharmaceutical ingredients benzoyl peroxide, t-butyl perbenzoate, and benzophenone. It is also used as an acylation reagent. Further, it is used in the preparation of peroxides such as benzoyl peroxide, benzophenone and t-butyl perbenzoate. In addition to this, it is involved in the preparation of chromones and antiproliferative agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Butyryl chloride is used as an intermediate for the synthesis of active pharmaceutical ingredients benzoyl peroxide, t-butyl perbenzoate, and benzophenone. It is also used as an acylation reagent. Further, it is used in the preparation of peroxides such as benzoyl peroxide, benzophenone and t-butyl perbenzoate. In addition to this, it is involved in the preparation of chromones and antiproliferative agents.

Solubility
Miscible with ether.

Notes
Moisture sensitive. Incompatible with water, alcohols, strong bases and strong oxidizing agents
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-75-3
Molecular Formula C4H7ClO
Molecular Weight (g/mol) 106.55
MDL Number MFCD00000752
InChI Key DVECBJCOGJRVPX-UHFFFAOYSA-N
Synonym butyryl chloride, n-butyryl chloride, butyric acid chloride, butyroyl chloride, butyric chloride, butyrylchloride, unii-2xvm8e16ir, ccris 5989, n-butyric acid chloride, n-butanoyl chloride
PubChem CID 8855
IUPAC Name butanoyl chloride
SMILES CCCC(Cl)=O

Specifications

Melting Point -89°C
Density 1.026
Boiling Point 101°C to 102°C
Flash Point 18°C (64°F)
Odor Acrid
Refractive Index 1.412
Quantity 250 g
UN Number UN2353
Beilstein 605395
Sensitivity Moisture sensitive
Merck Index 14,1598
Solubility Information Miscible with ether.
Formula Weight 106.55
Percent Purity 98%
Chemical Name or Material Butyryl chloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.