missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Consort™ C3010 Benchtop Multiparameter Analyzer
A full parameter dual channel instrument capable of performing 2 measurements simultaneously or individually shown on the display.
Brand: Consort™ C3010
Code : A0
Additional Details : Weight : 0.80000kg
Description
- No interference between pH/ORP/Ion and conductivity electrodes in the same solution
- Custom calibration tables allow the user to add complete buffer/standard-temperature relation tables
- Wide display shows 2 channels at the same time including temperature and date/time
- Stability indicator ensures visualization when the measurement has stabilized
- Stability algorithm ensures stable readings with ability to detect fast changes
- Hold function to freeze the display
- Selectable resolution for more stable readings of mV, pH and DO
- Range lock for conductivity measurements
- Capacitive compensation eliminates the capacitive component of the electrode and cable when measuring low conductivities
- Galvanic isolated USB interface eliminates ground looop effects when connected to a PC
- Pre-programmed standards for pH: 1.68, 2.00, 4.00, 4.01, 6.87, 6.99, 9.18, 9.21, 10.01, 12.00, 12.45 at 25°C
- Pre-programmed standards for conductivity: 1413 µS/cm, 12.88 mS/cm, 111.8 mS/cm at 25°C
- Free software and firmware updates downloadable from www.consort.be
Specifications
0.5% f.s. of range | |
0.1% ±1 digit | |
GLP, 0 to 999 hr reminder | |
LCD, 240 x 64 pixel | |
±2000 mV | |
0 to 200 MΩ cm | |
0 to 100 g/L | |
-5°C to +105°C | |
English, Dutch, French, German | |
600 to 1300 hPa | |
0.1 mV | |
0.01mg/L | |
26 cm | |
1 kg |
0.1% ± 1 digit | |
White back-lit | |
2 measurement, 2 temperature | |
0 to 2000 mS/cm | |
-2.000 to 16.000 | |
0.0 to 70.0 | |
0.001 | |
Measurement: 2 BNC, 1012 Ω Temperature: 2 x 2 bannanas, for Pt1000 |
|
Mains: 100 to 240 VAC, 50/60 Hz Low voltage: 9 to 15 VDC |
|
0.001 μS/cm | |
1 Ω cm | |
0.1°C | |
9 cm | |
240 VAC, 50/60 Hz |