missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CA2 (Human) Recombinant Protein
Human CA2 partial ORF (NP_000058.1, 191 a.a. - 260 a.a.) recombinant protein with GST tag at N-terminal.
335.00 € - 508.00 €
Specifications
Accession Number | NP_000058.1 |
---|---|
Gene ID (Entrez) | 760 |
Name | carbonic anhydrase II |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16033705
|
Abnova™
H00000760-Q01.10UG |
10 μg |
335.00 €
10µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16043705
|
Abnova™
H00000760-Q01.25UG |
25 μg |
508.00 €
25µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: WTYPGSLTTPPLLECVTWIVLKEPISVSSEQVLKFRKLNFNGEGEPEELMVDNWRPAQPLKNRQIKASFK
Specifications
NP_000058.1 | |
carbonic anhydrase II | |
125% SDS-PAGE Stained with Coomassie Blue | |
CA-II, CAII, Car2 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
760 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CA2 | |
GST |