Learn More
James Bedford Dewsbury™ Safety Storage Cabinet
Use these high quality cabinets to safely store flammable materials. James Bedford Dewsbury™ Safety Storage Cabinets feature robust welded construction in 0.9mm thick steel designed to maintain integrity for 30 minute fire rating.
Brand:  James Bedford Dewsbury™ 88F644G
Code : 03
Additional Details : Weight : 16.20000kg
Product Code. 12903415
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • With adjustable spillage and liquid tight sump tray
  • Doors are rebated and secured by chrome three way locking handle
  • Available in high visibility yellow or grey powder coated finish
  • Appropriate warning labels included
  • Cabinets compliant with:
  • D S E A R 2002
  • HS(G)51, 1990
  • Factory Inspectorate Certificate of Approval No 1: Parts 3 and 4
  • COSHH