Learn More
Cadence Science™ Deflected Noncoring Septum Penetration Needles
Use in gas chromatography and nuclear magnetic resonance spectroscopy procedures.
POA
Specifications
Material | Nickel-plated Brass (Hub), Stainless Steel (Tube) |
---|---|
For Use With (Application) | Use in gas chromatography and nuclear magnetic resonance spectroscopy procedures |
Includes | Deflected non-coring septum |
Product Type | Penetration Needle |
Needle Point Style | Deflected End |
Description
Use in gas chromatography and nuclear magnetic resonance spectroscopy procedures
- Penetration needles with deflected noncoring septum
- For noncoring penetration of septa and rubber closures involving air-sensitive and pyrophoric compounds
Specifications
Nickel-plated Brass (Hub), Stainless Steel (Tube) | |
Deflected non-coring septum | |
Deflected End |
Use in gas chromatography and nuclear magnetic resonance spectroscopy procedures | |
Penetration Needle |