missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Cadmium foil, 1.0mm (0.04 in.) thick, 15x15cm (5.9x5.9 in.), 99.95% (metals basis)

Brand:  Alfa Aesar™ 000940.KJ

Code : NEW

Additional Details : CAS Number : 7440-43-9 Weight : 0.01000kg

Quantity 5 pc.

 View more versions of this product

Product Code. 15414735

  • 1194.00 € / Pack of 5


Estimated Shipment date
from Supplier 31-08-2018
Add to basket

Specifications

Chemical Identifiers

7440-43-9
112.414
BDOSMKKIYDKNTQ-UHFFFAOYSA-N
CHEBI:22977
Cd
MFCD00010914
23973

Specifications

Cadmium foil
5 pc.
7440-43-9
(metals basis)
MFCD00010914
[Cd]
112.414
CHEBI:22977
≈195g/15x15cm
1.0mm (0.04 in.) thick, 15x15cm (5.9x5.9 in.)
99.95%
Cd
BDOSMKKIYDKNTQ-UHFFFAOYSA-N
cadmium
23973
Foil
Videos
SDS