missing translation for 'onlineSavingsMsg'
Learn More

Calcium carbonate, 99.999%, (trace metal basis), Thermo Scientific Chemicals

Catalog Number 11944311
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
471-34-1
CCaO3
100.09
MFCD00010906
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
10112
CHEBI:3311
[Ca++].[O-]C([O-])=O
This item is not returnable. View return policy
471-34-1
CCaO3
100.09
MFCD00010906
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
10112
CHEBI:3311
[Ca++].[O-]C([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY
Melting Point 825.0°C
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 39 to 42% (Complexometry)
Linear Formula CaCO3
Merck Index 15,1659
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in diluted acids
IUPAC Name calcium;carbonate
Formula Weight 100.09
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Calcium carbonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'