missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride, 93%, extra pure, powder, anhydrous, Thermo Scientific Chemicals

Catalog Number p-46072
12.93 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
500 g
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
10043-52-4
CaCl2
110.98
MFCD00010903
UXVMQQNJUSDDNG-UHFFFAOYSA-L
calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
5284359
CHEBI:3312
[Cl-].[Cl-].[Ca++]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
10043-52-4
CaCl2
110.98
MFCD00010903
UXVMQQNJUSDDNG-UHFFFAOYSA-L
calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
5284359
CHEBI:3312
[Cl-].[Cl-].[Ca++]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent
TRUSTED_SUSTAINABILITY
Boiling Point >1600.0°C
Melting Point 782.0°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 95% min. (Complexometry)
Linear Formula CaCl2
Fieser 01,104; 04,368
Merck Index 15,1661
Solubility Information Solubility in water: 740g/L (20°C). Other solubilities: freely soluble in alcohol
IUPAC Name calcium dichloride
Formula Weight 110.99
Percent Purity 96%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Calcium chloride, 96%
Show More Show Less
compliance-icons
Product Identifier
  • Calcium chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'