missing translation for 'onlineSavingsMsg'
Learn More

Calcium hydride, ca. 93%, extra pure, 10-100 mm pieces, up to 10% powder, Thermo Scientific Chemicals

Catalog Number p-46112
57.13 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7789-78-8
CaH2
42.09
MFCD00010897
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7789-78-8
CaH2
42.09
MFCD00010897

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent, Reduction reagent
TRUSTED_SUSTAINABILITY
Melting Point 675.0°C
Color Gray
Physical Form Irregular Lumps/Pieces
Quantity 100 g
Assay Percent Range 92% min. (Total Ca)
Linear Formula CaH2
Fieser 01,105
Merck Index 15,1674
Solubility Information Solubility in water: reacts. Other solubilities: decomposes with lower alcohols and carboxylic acid, soluble only in salt melts
Formula Weight 42.09
Percent Purity ∼93%
Grade Extra Pure
Packaging Plastic bottle
Specific Gravity 1.9
Density 1.9000g/mL
Chemical Name or Material Calcium hydride, ca. 93%
Show More Show Less
compliance-icons
Product Identifier
  • Calcium hydride
Signal Word
  • Danger
Hazard Category
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
Precautionary Statement
  • P223-Do not allow contact with water.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.
  • P370+P378-In case of fire: Use to extinguish.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'