Learn More
Calcium Nitrate Tetrahydrate, Extra Pure, SLR, Fisher Chemical™
Brand: Fisher Chemical C/1880/53
Code : 21
Additional Details : CAS Number : 10124-37-5 Weight : 0.50000kg
Specifications
13477-34-4 | |
149604 | |
Solid Crystalline | |
132°C | |
45°C | |
500g |
CaN2O6·4H2O | |
236.15g/mol | |
Plastic powder jar | |
White | |
5 to 7 |