Learn More
Thermo Scientific™ Capitol Vial Flip-Top Polypropylene Containers with Lock Seal
Capitol Vial flip-top vials with lock seal prevent accidental opening during handling, transport and storage.
99.90 € - 320.00 €
Product Code | Brand | Volume (English) | Material | Color | Diameter (Metric) Inner | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (English) | Material | Color | Diameter (Metric) Inner | Price | Quantity & Availability | ||
15688508
|
Thermo Scientific™
02LS |
1.5 oz. | Polypropylene | Clear | 28 mm |
227.00 €
Pack of 600 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
||
15698508
|
Thermo Scientific™
02LSGREEN |
1.5 oz. | Polypropylene | Green | 28 mm |
320.00 €
Pack of 600 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
||
17155261
|
Thermo Scientific™
0385BLACK |
3 oz. | Polypropylene | Black | 33 mm |
202.00 €
Pack of 600 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
||
15688518
|
Thermo Scientific™
0385YELLOW |
3 oz. | Polypropylene | Yellow | 33 mm |
202.00 €
Pack of 400 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
||
15678538
|
Thermo Scientific™
04HPLS |
4 oz. | Polypropylene | Natural | 43 mm |
99.90 €
Pack of 250 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
||
17105281
|
Thermo Scientific™
04HPLS ROYALBLUE |
4 oz. | Polypropylene | Royal Blue |
105.00 €
Pack of 250 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||
15698538
|
Thermo Scientific™
04LPLS |
4 oz. | Polypropylene | Natural | 63mm |
224.00 €
Pack of 250 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
||
15608548
|
Thermo Scientific™
04LPLSBLACK |
4 oz. | Polypropylene | Natural | 63 mm |
267.00 €
Pack of 250 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||
15618548
|
Thermo Scientific™
04LPLSGOLD |
5 oz. | Polypropylene | Natural | 63 mm |
219.00 €
Pack of 250 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
||
17135281
|
Thermo Scientific™
04LPLSRED |
6 oz. | Polypropylene | Natural | 63 mm |
219.00 €
Pack of 250 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
||
17155281
|
Thermo Scientific™
04LPLSYELLOW |
7 oz. | Polypropylene | Natural | 63 mm |
235.00 €
Pack of 250 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
||
15658548
|
Thermo Scientific™
08LS |
8 oz. | Polypropylene | Natural | 75 mm |
152.00 €
Pack of 140 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
||
15628568
|
Thermo Scientific™
10LSCL |
10 oz. | Polypropylene | Natural | 62 mm |
107.00 €
Pack of 100 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
||
17115291
|
Thermo Scientific™
10LSRED |
10 oz. | Polypropylene | Red | 62 mm |
145.00 €
Pack of 250 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
||
15638568
|
Thermo Scientific™
10LSWHITE |
10 oz. | Polypropylene | White | 62 mm |
145.00 €
Pack of 100 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
||
15648568
|
Thermo Scientific™
10LSYELLOW |
10 oz. | Polypropylene | Yellow | 62 mm |
145.00 €
Pack of 100 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
||
15618568
|
Thermo Scientific™
10LS BLACK |
10 oz. | Polypropylene | Black | 62 mm |
145.00 €
Pack of 100 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
||
Description
Thermo Scientific™ Capitol Vial Flip-Top Polypropylene Containers with Lock Seal feature a locking tab on the lid to prevent accidental opening during handling and transport.
Thermo Scientific™ Capitol Vial Flip-Top Polypropylene Containers with Lock Seal are ideal for collecting, transporting and storing a wide range of liquids and solids.
- Constructed from food grade polypropylene
- Airtight and leak-resistant
- Permanent hinged lid for easy opening with one hand
- Non-Sterile
- Available in capacities ranging from 1.5 to 10 oz. (45 to 296 mL)
Recommended for:
Food and beverage testing, veterinary applications, environmental sampling, general packaging and storage