missing translation for 'onlineSavingsMsg'
Learn More

Carbon Standard For ICP, 1000μg/mL in H2O, Assurance Grade, SPEX CertiPrep™

Catalog Number p-5533048
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Triple-leached LDPE bottle
This item is not returnable. View return policy
7440-44-0, 7732-18-5
C
12.011
OKTJSMMVPCPJKN-UHFFFAOYSA-N
5462310
CHEBI:27594
carbon
[C]
This item is not returnable. View return policy
7440-44-0, 7732-18-5
C
12.011
OKTJSMMVPCPJKN-UHFFFAOYSA-N
5462310
CHEBI:27594
carbon
[C]

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
C1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

CAS Min % 0.1, 99.9
CAS Max % 0.1, 99.9
Grade Assurance
Concentration or Composition (by Analyte or Components) Matrix Concentration Cat. No.
Oxalic Acid in H2O 1000μg/mL PLC9-2Y, -2X 
Packaging Triple-leached LDPE bottle
Certificate of Analysis Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
Solvent or Matrix H2O
Quantity 125 mL
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Chemical Name or Material Carbon Standard
Show More Show Less

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'