Learn More
Invitrogen™ Carbonic Anhydrase XIII Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5120274
Product Code. 17741674
  • 504.00 € / 100µL
Description
Positive test controls include: HT-29, DU145, Mouse spleen, Mouse lung, Mouse kidney, Mouse heart. Immunogen sequence: MSRLSWGYRE HNGPIHWKEF FPIADGDQQS PIEIKTKEVK YDSSLRPLSI KYDPSSAKII SNSGHSFNVD FDDTENKSVL RGGPLTGSYR LRQVHLHWGS ADDHGSEHIV DGVSYAAELH VVHWNSDKYP SFVEAAHEPD GLAVLGVFLQ IGEPNSQLQK ITDTLDSIKE KGKQTRFTNF DLLSLLPPSW DYWTYPGSLT VPPLLESVTW IVLKQPINIS.
Carbonic anhydrase XII (CA XII), encoded by the CA12 gene, is one of the enzyme isoforms in the carbonic anhydrase family that catalyzes the reversible hydration of carbon dioxide into bicarbonate and protons. CA XII is a transmembrane protein expressed in various tissues, including the kidneys, intestines, and lungs, playing crucial roles in maintaining acid-base balance, pH regulation, and electrolyte transport. Its activity is significant in organs where fluid secretion and absorption are essential processes, impacting functions such as respiration and renal bicarbonate reabsorption. CA XII is involved in physiological processes like cell proliferation and differentiation by modulating the extracellular and intracellular pH, influencing tumor microenvironment dynamics. Aberrant expression of CA XII has been noted in several cancers, including breast and colorectal cancers, where it supports tumor growth and metastasis by maintaining the acid-base homeostasis conducive to cancer cell survival. Due to its surface expression and role in maintaining pH balance, CA XII is a target of interest for therapeutic interventions aiming to disrupt tumor progression and enhance the efficacy of cancer treatments.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Carbonic Anhydrase XIII
Polyclonal
Unconjugated
Ca13
2310075C21Rik; Ca13; Car13; Carbonate dehydratase XIII; carbonic anhydrase 13; Carbonic anhydrase XIII; CAXIII; CA-XIII; RGD1560453
Rabbit
Affinity Chromatography
RUO
377677, 71934
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot
1.08 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Q8N1Q1, Q9D6N1
Ca13, Car13
Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human CA13 (NP_940986.1).
100 μL
Primary
Human, Mouse