Learn More
Fisherbrand™ Cardboard Cryoboxes, 136mm
Combine these boxes with different grid inserts to hold a wide range of tube sizes. Fisherbrand™ Cardboard Cryoboxes are temperature resistant down to -80°C, ideal for ultra-low temperature storage.
34.35 € - 179.00 €
Specifications
Material Cardboard
Type Cryo Box
Quantity 10 Pack
Dimensions (L x W) 136 x 136 mm
Length (Metric) 136 mm
View More Specs
Products 22
Product Code Brand Color Dimensions (L x W x H) Price Quantity & Availability  
Product Code Brand Color Dimensions (L x W x H) Price Quantity & Availability  
11822463
View accessory products View Documents Promotion Details
Fisherbrand™
Red 136 x 136 x 100 mm
79.10 €
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 05-06-2024
to see stock.
Add to basket
11822453
View accessory products View Documents Promotion Details
Fisherbrand™
Red 136 x 136 x 50 mm
37.25 €
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Add to basket
11872453
View accessory products View Documents Promotion Details
Fisherbrand™
Red 136 x 136 x 75 mm
46.85 €
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 05-07-2024
to see stock.
Add to basket
11842453
View accessory products View Documents Promotion Details
Fisherbrand™
Yellow 136 x 136 x 50 mm
38.20 €
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 29-04-2024
to see stock.
Add to basket
11557633
View accessory products View Documents Promotion Details
Fisherbrand™
White 136 x 136 x 100 mm
179.00 €
Pack of 10
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
11852463
View accessory products View Documents Promotion Details
Fisherbrand™
White 136 x 136 x 130 mm
72.50 €
Pack of 10
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
11862443
View accessory products View Documents Promotion Details
Fisherbrand™
Red 136 x 136 x 32 mm
59.00 €
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 05-07-2024
to see stock.
Add to basket
11862463
View accessory products View Documents Promotion Details
Fisherbrand™
Blue 136 x 136 x 130 mm
70.25 €
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Add to basket
11852443
View accessory products View Documents Promotion Details
Fisherbrand™
Blue 136 x 136 x 32 mm
59.00 €
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 05-07-2024
to see stock.
Add to basket
11812463
View accessory products View Documents Promotion Details
Fisherbrand™
Blue 136 x 136 x 100 mm
79.10 €
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 05-06-2024
to see stock.
Add to basket
11527643
View accessory products View Documents Promotion Details
Fisherbrand™
Red 136 x 136 x 130 mm
70.25 €
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 05-07-2024
to see stock.
Add to basket
11507623
View accessory products View Documents Promotion Details
Fisherbrand™
White 136 x 136 x 32 mm
34.35 €
Pack of 10
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
11507633
View accessory products View Documents Promotion Details
Fisherbrand™
White 136 x 136 x 75 mm
89.20 €
Pack of 10
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
11812453
View accessory products View Documents Promotion Details
Fisherbrand™
Blue 136 x 136 x 50 mm
37.25 €
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 17-04-2024
to see stock.
Add to basket
11812473
View accessory products View Documents Promotion Details
Fisherbrand™
Yellow 136 x 136 x 130 mm
70.25 €
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 05-07-2024
to see stock.
Add to basket
11547623
View accessory products View Documents Promotion Details
Fisherbrand™
Yellow 136 x 136 x 32 mm
59.00 €
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 05-06-2024
to see stock.
Add to basket
11882453
View accessory products View Documents Promotion Details
Fisherbrand™
Green 136 x 136 x 75 mm
46.85 €
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 05-07-2024
to see stock.
Add to basket
11872443
View accessory products View Documents Promotion Details
Fisherbrand™
Green 136 x 136 x 32 mm
66.15 €
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 05-07-2024
to see stock.
Add to basket
11587623
View accessory products View Documents Promotion Details
Fisherbrand™
Green 136 x 136 x 50 mm
39.45 €
Pack of 10
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
11842463
View accessory products View Documents Promotion Details
Fisherbrand™
Yellow 136 x 136 x 100 mm
79.10 €
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 05-07-2024
to see stock.
Add to basket
11802473
View accessory products View Documents Promotion Details
Fisherbrand™
Green 136 x 136 x 130 mm
70.25 €
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 05-07-2024
to see stock.
Add to basket
11832463
View accessory products View Documents Promotion Details
Fisherbrand™
Green 136 x 136 x 100 mm
81.50 €
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 05-07-2024
to see stock.
Add to basket
Description
Description
  • Available with a water-repellent finish or with a waterproof plastic coating
  • Highest flexibility: Cryoboxes can be combined with different grid inserts for tubes from 6 to 28.5mm in diameter
  • The Cryoboxes can be used for larger vessels if the grid is removed
Specifications
Specifications
Cardboard
10 Pack
136 mm
Cryo Box
136 x 136 mm
136 mm