Learn More
Thermo Scientific™ Dionex™ SolEx™ Cartridge Holder V-3
Order this holder when using the Thermo Scientific™ Dionex™ SolEx™ HRP cartridge for online SPE preconcentration of water samples prior to HPLC.
Brand: Thermo Scientific™ 074403
Code : 16
Additional Details : Weight : 0.50000kg
Description
Required when using the cartridge format of Thermo Scientific™ Dionex™ SolEx™ HRP columns. The Dionex SolEx Cartridge Holder V-3 is designed to make it easy to change cartridges when necessary. Dionex SolEx HRP columns and cartridges provide fast, easy sample pre-concentration of contaminants from water samples by solid-phase extraction prior to HPLC analysis. Used online, they are placed in the valve to automatically concentrate the analytes of interest.
Specifications
V-3 Holder | |
On-Line SPE Concentration Columns | |
SolEx | |
V-3 Cartridge Folder |