Learn More
Emma Safety Footwear CAS Safety Shoes
Washable microfiber uppers are treated to repel all kinds of dirt, germs and bacteria. A highly breathable, comfortable shoe for all day wear. Energy returning soles prevent fatigue and keep the wearer fresh, alert, foot sure and steady even in the event of a spillage.
Brand:  Emma Safety Footwear MM52054041
Additional Details : Weight : 1.37900kg
 View more versions of this product
Product Code. 15918671
  • 108.12 € / 1 pair
Estimated Shipment: 24-09-2024
to see stock.
Add to basket
Description
Description
  • S2-Safety shoe for wet working conditions
  • GripForce™ Easy Twist Outsole
  • Microfiber construction
  • Chrome-free lining
  • Hydro-Tec™ Sanitized Silver
Specifications
Specifications
EN ISO 20345:2011
White
Hydro-Tec™ Sanitized Silver
41
Laces
CAS
Microfiber
Steel