missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Cassette

For use with the Fraction collector F9-C and the ÄKTA avant chromatography system. Cytiva Cassettes are available for Deep-Well Plates or 3mL Tubes for large-scale processing of samples in chromatography applications. Pack of 2.

Brand:  Cytiva 28-9564-27

Code : GE

Additional Details : Weight : 0.73600kg

 View more versions of this product

Product Code. 11949155

  • 125.00 € / Pack of 2


Estimated Shipment
28-04-2021
Add to basket

Description

Description

Cytiva Cassettes are available in packs of two (2) for large-scale processing of samples in chromatography applications using the Fraction collector F9-C and the ÄKTA avant chromatography system.
  • For Deep-Well Plates (in Flexible fraction collector): Holds 24, 48 or 96 deep-well plate
  • For 3mL Tubes: Holds up to 40 tubes
Specifications

Specifications

Chromatography Applications
Cassette
2/Pk.
Fraction Collector F9-C and ÄKTA Avant Chromatography System
For Use With 3 mL tubes
Videos
SDS
Documents