missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Castor oil, specified according to the requirements of Ph.Eur.

Catalog Number 11974041
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
8001-79-4
C57H104O9
933.45
MFCD00130746
ZEMPKEQAKRGZGQ-AAKVHIHISA-N
castor oil, ricinus oil, olio di ricino, venelex, xenaderm, optase, trypsin complex, unii-d5340y2i9g, castor oil usp:jan, 1-o,2-o,3-o-tris z-12-hydroxy-9-octadecenoyl glycerol
14030006
2,3-bis[[(Z)-12-hydroxyoctadec-9-enoyl]oxy]propyl (Z)-12-hydroxyoctadec-9-enoate
CCCCCCC(CC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCC(CCCCCC)O)OC(=O)CCCCCCCC=CCC(CCCCCC)O)O
This item is not returnable. View return policy
8001-79-4
C57H104O9
933.45
MFCD00130746
ZEMPKEQAKRGZGQ-AAKVHIHISA-N
castor oil, ricinus oil, olio di ricino, venelex, xenaderm, optase, trypsin complex, unii-d5340y2i9g, castor oil usp:jan, 1-o,2-o,3-o-tris z-12-hydroxy-9-octadecenoyl glycerol
14030006
2,3-bis[[(Z)-12-hydroxyoctadec-9-enoyl]oxy]propyl (Z)-12-hydroxyoctadec-9-enoate
CCCCCCC(CC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCC(CCCCCC)O)OC(=O)CCCCCCCC=CCC(CCCCCC)O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Acidity 2.0mg KOH/g max.
Absorbance 1.5 max. (at the absorption maximum at 269 +/- 1nm)
Melting Point -10.0°C
Color Colorless or Yellow
Density 0.9570g/mL
Boiling Point 229.0°C
Flash Point 229°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4770 to 1.4810
Quantity 2.5 L
Merck Index 14,1898
Specific Gravity 0.957
Specific Rotation Condition + 3.50 (20.00°C c=neat)
Specific Rotation + 3.50
Solubility Information Solubility in water: .. Other solubilities: miscible with alcohol, benzene, chloroform,, carbondisulfide, ether and glacial acetic acid
Viscosity 0,6-0,8 Pas (25°C)
Grade Eu. Ph.
Concentration or Composition (by Analyte or Components) of fatty acids (GC): Passes Test (per Eur. Pharm.)
Physical Form Viscous Hygroscopic Liquid
Chemical Name or Material Castor oil
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'