missing translation for 'onlineSavingsMsg'
Learn More

Catechol, Extra Pure, SLR, Fisher Chemical™

Catalog Number p-7072912
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
120-80-9
C6H6O2
2188
This item is not returnable. View return policy
120-80-9
C6H6O2
2188

Catechol, Extra Pure, SLR

TRUSTED_SUSTAINABILITY
Melting Point 103°C
Color Brown
pH 6
Boiling Point 245°C
Packaging Plastic powder jar
Quantity 100 g
Formula Weight 110.11g/mol
Physical Form Solid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'