missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Catenin, gamma Mouse, Clone: CTNG/1483, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 3728-MSM3-P1
Additional Details : Weight : 0.01000kg
Specifications
Catenin, gamma | |
Cell Surface and Cytoplasmic | |
CTNG/1483 | |
Unconjugated | |
NP_001339702 | |
Mouse | |
80-87kDa | |
Protein A/G Purified | |
RUO | |
3728 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
JUP | |
Recombinant human full-length gamma catenin protein | |
A431, T47D or MCF-7 cells. Skin | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |