missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Catenin, gamma Mouse, Clone: CTNG/1664, enQuireBio™
Mouse Monoclonal Antibody
199.00 € - 485.00 €
Specifications
Antigen | Catenin, gamma |
---|---|
Clone | CTNG/1664 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15960308
|
EnquireBio
3728-MSM4-P1 |
100 μg |
485.00 €
100µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15950308
|
EnquireBio
3728-MSM4-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Catenin, gamma | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
3728 | |
Recombinant human full-length gamma catenin protein | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
80-87kDa |
CTNG/1664 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_001339702 | |
JUP | |
IgG1 κ | |
Protein A/G Purified | |
Cell Surface and Cytoplasmic | |
A431, T47D or MCF-7 cells. Skin |