Learn More
Cathepsin K Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11239-1-AP-150UL
 View more versions of this product
Product Code. 16828183
  • 410.00 € / 150µL
Description
The protein encoded by this gene is a lysosomal cysteine proteinase involved in bone remodeling and resorption. This protein, which is a member of the peptidase C1 protein family, is predomitly expressed in osteoclasts. However, the encoded protein is also expressed in a significant fraction of human breast cancers, where it could contribute to tumor invasiveness. Mutations in this gene are the cause of pycnodysostosis, an autosomal recessive disease characterized by osteosclerosis and short stature. This gene may be subject to RNA editing.
Specifications
Specifications
Cathepsin K
Polyclonal
Unconjugated
CTSK
Cathepsin K, Cathepsin O, Cathepsin O2, Cathepsin X, CTS02, CTSK, CTSO, CTSO1, CTSO2, PKND, PYCD
Rabbit
Antigen Affinity Chromatography
RUO
13038, 1513, 29175
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O35186, P43235, P55097
Ctsk
Cathepsin K Fusion Protein Ag1761
150 μL
Primary
Human, Mouse, Rat