Learn More
Paragon Scientific™ CCS Viscosity Standards
Paragon ISO 17025 / ISO 17034 certified CCS viscosity standards are for the calibration and verification of analytical equipment used in Cold Cranking Simulator (CCS) testing to ASTM D5293 and SAE Specification J300.
66.21 € - 326.00 €
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
15568854
|
Paragon Scientific
CL090 |
SAE Specification J303 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15578854
|
Paragon Scientific
CL120 |
SAE Specification J306 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15588854
|
Paragon Scientific
CL140 |
SAE Specification J308 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15598854
|
Paragon Scientific
CL150 |
SAE Specification J309 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15508864
|
Paragon Scientific
CL160 |
SAE Specification J310 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15518864
|
Paragon Scientific
CL170 |
SAE Specification J311 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15528864
|
Paragon Scientific
CL190 |
SAE Specification J312 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15538864
|
Paragon Scientific
CL220 |
SAE Specification J314 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15548864
|
Paragon Scientific
CL240 |
SAE Specification J315 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15558864
|
Paragon Scientific
CL250 |
SAE Specification J316 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15568864
|
Paragon Scientific
CL260 |
SAE Specification J317 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15578864
|
Paragon Scientific
CL280 |
SAE Specification J318 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15588864
|
Paragon Scientific
CL300 |
SAE Specification J319 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15598864
|
Paragon Scientific
CL320 |
SAE Specification J320 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15508874
|
Paragon Scientific
CL340 |
SAE Specification J321 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15518874
|
Paragon Scientific
CL380 |
SAE Specification J322 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15528874
|
Paragon Scientific
CL420 |
SAE Specification J323 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15538874
|
Paragon Scientific
CL480 |
SAE Specification J324 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15548874
|
Paragon Scientific
CL530 |
SAE Specification J325 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15558874
|
Paragon Scientific
CL600 |
SAE Specification J326 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15523556
|
Paragon Scientific
CL740 |
SAE Specification J328 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
- Full Traceability to National Standards
- Ensures full compliance to ASTM & IP test method protocol
- Manufactured in the United Kingdom
- 2 year shelf life
Nominal values quoted