missing translation for 'onlineSavingsMsg'
Learn More

CD11b Mouse, PE, Clone: ICRF44, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB25-PE-25Tests

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15991728

  • 95.36 € / 25 tests

Estimated Shipment: 03-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD11b
ICRF44
PE
XP_006721108
Mouse
Protein A/G Purified
RUO
Primary
3684
Flow Cytometry
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
ITGAM
IgG1 kappa
25 Tests
Store at 2-4°C.
Monoclonal
Videos
SDS
Documents
Certificates