Learn More
CD11c Armenian Hamster, APC-Cy7, Clone: N418, enQuireBio™
Armenian Hamster Monoclonal Antibody
Brand:  EnquireBio QAB23-APC7-25ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15991688
  • 95.36 € / 25µg
Estimated Shipment: 03-12-2021
Specifications
CD11c
N418
APC-Cyanine7
NP_067309
Armenian Hamster
Protein A/G Purified
RUO
Primary
16411
Flow Cytometry
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
Itgax
IgG
25 μg
Store at 2-4°C.