missing translation for 'onlineSavingsMsg'
Learn More

CD11c Mouse, APC, Clone: 3.9, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB24-APC-25Tests

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15991708

  • 95.36 € / 25 tests

Estimated Shipment: 25-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD11c
3.9
APC
XP_011544154
Mouse
Protein A/G Purified
RUO
Primary
3687
Flow Cytometry
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
ITGAX
IgG1 kappa
25 Tests
Store at 2-4°C.
Monoclonal
Videos
SDS
Documents
Certificates