missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11c Mouse, Clone: ITGAX/1284, enQuireBio™
Mouse Monoclonal Antibody
199.00 € - 485.00 €
Specifications
Antigen | CD11c |
---|---|
Clone | ITGAX/1284 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15990288
|
EnquireBio
3687-MSM4-P1 |
100 μg |
485.00 €
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15980288
|
EnquireBio
3687-MSM4-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
CD11c | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
3687 | |
Recombinant human ITGAX protein fragment (aa 637-827) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
145kDa |
ITGAX/1284 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
XP_011544154 | |
ITGAX | |
IgG2b | |
Protein A/G Purified | |
Cell surface | |
Human dendritic cells. Human lymph nodes and tonsils. |