missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD15 / FUT4 Rabbit, Clone: FUT4/1478R, enQuireBio™
Rabbit Monoclonal Antibody
Brand: EnquireBio 2526-RBM8-P1
Additional Details : Weight : 0.01000kg
Specifications
CD15 / FUT4 | |
Cell surface and granular paranuclear | |
FUT4/1478R | |
Unconjugated | |
NP_002024 | |
Rabbit | |
∽220kDa | |
Protein A/G Purified | |
RUO | |
2526 | |
IgG κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
FUT4 | |
Purified human neutrophils | |
U937 cells, Reed-Sternberg's cells in Hodgkin's lymphoma | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |