missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD161 Mouse, Biotin, Clone: PK136, enQuireBio™
Mouse Monoclonal Antibody
82.15 € - 276.00 €
Specifications
Antigen | CD161 |
---|---|
Clone | PK136 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15932478
|
EnquireBio
QAB78-B-25UG |
25 μg |
82.15 €
25µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15922478
|
EnquireBio
QAB78-B-100UG |
100 μg |
179.00 €
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15942478
|
EnquireBio
QAB78-B-500UG |
500 μg |
276.00 €
500µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD161 | |
0.5mg/mL | |
Monoclonal | |
Mouse | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
17059 | |
IgG2a κ | |
Protein A/G Purified |
PK136 | |
Flow Cytometry | |
Biotin | |
RUO | |
NP_001153376 | |
Klrb1c | |
Primary | |
Store at 2-4°C. |