missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD162 Recombinant Rabbit Monoclonal Antibody (001), APC

Rabbit Recombinant Monoclonal Antibody
39.00 € - 99.00 €
Specifications
| Antigen | CD162 |
|---|---|
| Clone | 001 |
| Concentration | 0.1 mg/mL |
| Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
| Applications | Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30533874
![]() |
Invitrogen™
MA560483 |
100 Tests |
99.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30534903
|
Invitrogen™
MA560484 |
25 Tests |
39.00 €
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD162 Recombinant Monoclonal Antibody for Flow
CD162 (PSGL-1) is a 120 kDa homodimer protein found on the surface of leukocytes and platelets. CD162 is known to bind CD62P (P-selectin), CD62L (L-selectin), and to a lower degree CD62E (E-selectin). These interactions play a critical role in leukocyte adhesion, rolling and extravasation. CD162 has been shown to be an important immune marker of T cell exhaustion in chronic viral infections and cancer.Specifications
| CD162 | |
| 0.1 mg/mL | |
| Flow Cytometry | |
| APC | |
| Rabbit | |
| RUO | |
| PBS with 0.5% BSA and 0.09% sodium azide | |
| Q62170 | |
| 20345 | |
| Recombinant Mouse PSGL-1/CD162 protein (Met1-Cys307). | |
| Antibody |
| 001 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Mouse | |
| SELPLG | |
| CD162; CLA; Cutaneous lymphocyte associated antigen; cutaneous lymphocyte-associated associated antigen; leukocyte cell surface adhesion molecule; P-selectin glycoprotein ligand 1; P-selectin glycoprotein ligand 1 propeptide; P-selectin glycoprotein ligand-1; Psgl1; Psgl-1; Selectin P ligand; selectin, platelet (p-selectin) ligand; Selp1; SELPG; Selpl; SELPLG | |
| SELPLG | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title