missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD178 / Fas ligand Mouse, APC, Clone: NOK-1, enQuireBio™
Mouse Monoclonal Antibody
143.00 € - 288.00 €
Specifications
Antigen | CD178 / Fas ligand |
---|---|
Clone | NOK-1 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15932568
|
EnquireBio
QAB85-APC-25TESTS |
25 Tests |
143.00 €
25 tests |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15922568
|
EnquireBio
QAB85-APC-100TESTS |
100 Tests |
288.00 €
100 tests |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD178 / Fas ligand | |
0.5mg/mL | |
Monoclonal | |
Mouse | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
356 | |
IgG1 κ | |
Protein A/G Purified |
NOK-1 | |
Flow Cytometry | |
APC | |
RUO | |
NP_001289675 | |
FASLG | |
Primary | |
Store at 2-4°C. |