missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences CD19 Isolation Kits for FABian™, Human
IBA Lifesciences CD19 Isolation Kits for FABian™, Human for reversible cell isolation delivering label-free target cells.
90.41 € - 718.00 €
Specifications
Product Type | Cell Isolation Kit |
---|---|
Content And Storage | 2°C to 8°C |
For Use With (Application) | Selection of CD19+ Cells From Whole Blood or Buffy Coat in Conjunction with The FABian™ System |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16461485
|
IBA Lifesciences
6-6013-001 |
1 Kit |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16471485
|
IBA Lifesciences
6-6013-010 |
10 Kits |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- The kit is used for the selection of CD19+ cells from whole blood or buffy coat in conjunction with the FABian™ system
- It includes CD19 Fab-Strep (human), the Cell Isolation Column, Cell Isolation Buffer, Biotin for Cell Elution and tubes and a luer lock adapter with the syringe
- Each kit is sufficient for one FABian™ run to isolate cells from 9 ml whole blood or 6.4 mL buffy coat
Specifications
Cell Isolation Kit | |
2°C to 8°C | |
Selection of CD19+ Cells From Whole Blood or Buffy Coat in Conjunction with The FABian™ System |