missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Mouse anti-Human, Clone: CAT-13.1E10, Novus Biologicals™
Mouse Monoclonal Antibody
266.00 €
Specifications
| Antigen | CD19 |
|---|---|
| Clone | CAT-13.1E10 |
| Concentration | 1 mg/ml |
| Dilution | Flow Cytometry, Immunocytochemistry/Immunofluorescence |
| Applications | Flow Cytometry, Immunofluorescence |
Description
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
| CD19 | |
| 1 mg/ml | |
| Flow Cytometry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Lines, Stem Cell Markers | |
| PBS | |
| 930 | |
| IgG1 κ | |
| Protein A purified |
| CAT-13.1E10 | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12 | |
| This antibody was raised by immunising mice against CD19. | |
| Primary | |
| Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts