missing translation for 'onlineSavingsMsg'
Learn More

CD19 Mouse, PE, Clone: SJ25C1, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB29-PE-500Tests

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15931788

  • 915.00 € / 500 tests
Estimated Shipment: 19-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD19
Monoclonal
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
CD19
Protein A/G Purified
RUO
930
IgG1 κ
Flow Cytometry
SJ25C1
PE
NP_001171569
Mouse
500 Tests
Primary
Store at 2-4°C.
Videos
SDS
Documents
Certificates
Special Offers

Special Offers