missing translation for 'onlineSavingsMsg'
Learn More

CD19 Mouse, PerCP, Clone: SJ25C1, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB29-PCP-25Tests

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15991778

  • 170.61 € / 25 tests

Estimated Shipment: 25-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD19
SJ25C1
PerCP
NP_001171569
Mouse
Protein A/G Purified
RUO
Primary
930
Flow Cytometry
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
CD19
IgG1 kappa
25 Tests
Store at 2-4°C.
Monoclonal
Videos
SDS
Documents
Certificates