missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD20 Mouse, FITC, Clone: 2H7, enQuireBio™
Mouse Monoclonal Antibody
131.00 € - 1295.00 €
Specifications
Antigen | CD20 |
---|---|
Clone | 2H7 |
Concentration | 5μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15971828
|
EnquireBio
QAB31-F-25TESTS |
25 Tests |
131.00 €
25 tests |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15961828
|
EnquireBio
QAB31-F-100TESTS |
100 Tests |
288.00 €
100 tests |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15981828
|
EnquireBio
QAB31-F-500TESTS |
500 Tests |
1295.00 €
500 tests |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD20 | |
5μL/Test | |
Monoclonal | |
Mouse | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
931 | |
IgG2b κ | |
Protein A/G Purified |
2H7 | |
Flow Cytometry | |
FITC | |
RUO | |
NP_690605 | |
MS4A1 | |
Primary | |
Store at 2-4°C. |