missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD25 Mouse, PE-Cy7, Clone: BC96, enQuireBio™
Mouse Monoclonal Antibody
155.00 € - 312.00 €
Specifications
Antigen | CD25 |
---|---|
Clone | BC96 |
Concentration | 5μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15901888
|
EnquireBio
QAB35-PE7-25TESTS |
25 Tests |
155.00 €
25 tests |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15991878
|
EnquireBio
QAB35-PE7-100TESTS |
100 Tests |
312.00 €
100 tests |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD25 | |
5μL/Test | |
Monoclonal | |
Mouse | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
3559 | |
IgG1 κ | |
Protein A/G Purified |
BC96 | |
Flow Cytometry | |
PE-Cyanine7 | |
RUO | |
NP_001295171 | |
IL2RA | |
Primary | |
Store at 2-4°C. |