missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD28 Mouse, FITC, Clone: CD28.2, enQuireBio™
Mouse Monoclonal Antibody
94.25 € - 203.00 €
Specifications
Antigen | CD28 |
---|---|
Clone | CD28.2 |
Concentration | 5μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15901908
|
EnquireBio
QAB37-F-25TESTS |
25 Tests |
94.25 €
25 tests |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15991898
|
EnquireBio
QAB37-F-100TESTS |
100 Tests |
203.00 €
100 tests |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD28 | |
5μL/Test | |
Monoclonal | |
Mouse | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
940 | |
IgG1 κ | |
Protein A/G Purified |
CD28.2 | |
Flow Cytometry | |
FITC | |
RUO | |
NP_006130 | |
CD28 | |
Primary | |
Store at 2-4°C. |