missing translation for 'onlineSavingsMsg'
Learn More

CD28 Mouse, FITC, Clone: CD28.2, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB37-F-100Tests

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15991898

  • 170.61 € / 100 tests

Estimated Shipment: 30-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD28
CD28.2
FITC
NP_006130
Mouse
Protein A/G Purified
RUO
Primary
940
Flow Cytometry
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
CD28
IgG1 kappa
100 Tests
Store at 2-4°C.
Monoclonal
Videos
SDS
Documents
Certificates