missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Mouse, PE, Clone: UCHT1, enQuireBio™
Mouse Monoclonal Antibody
82.15 € - 615.00 €
Spezifikation
Antigen | CD3 |
---|---|
Clone | UCHT1 |
Concentration | 5μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
15942258
|
EnquireBio
QAB6-PE-25TESTS |
25 Tests |
82.15 €
25 tests |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15932258
|
EnquireBio
QAB6-PE-100TESTS |
100 Tests |
216.00 €
100 tests |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15952258
|
EnquireBio
QAB6-PE-500TESTS |
500 Tests |
615.00 €
500 tests |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Spezifikation
CD3 | |
5μL/Test | |
Monoclonal | |
Mouse | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
915 | |
IgG1 κ | |
Protein A/G Purified |
UCHT1 | |
Flow Cytometry | |
PE | |
RUO | |
NP_001035741 | |
CD3D | |
Primary | |
Store at 2-4°C. |