missing translation for 'onlineSavingsMsg'
Learn More

CD3 Mouse, PerCP-Cy5.5, Clone: OKT3, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB5-PCP55-100Tests

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15992128

  • 287.45 € / 100 tests

Estimated Shipment: 30-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD3
OKT3
PerCP-Cyanine5.5
NP_000724
Mouse
Protein A/G Purified
RUO
Primary
916
Flow Cytometry
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
CD3E
IgG2a kappa
100 Tests
Store at 2-4°C.
Monoclonal
Videos
SDS
Documents
Certificates