missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Rat, PE, Clone: 17A2, enQuireBio™
Rat Monoclonal Antibody
Brand: EnquireBio QAB3-PE-100ug
Additional Details : Weight : 0.01000kg
Specifications
CD3 | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Cd3g | |
Protein A/G Purified | |
RUO | |
12502 | |
IgG2b κ |
Flow Cytometry | |
17A2 | |
PE | |
NP_033980 | |
Rat | |
100 μg | |
Primary | |
Store at 2-4°C. |