missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD30 / TNFRSF8 Rabbit, Clone: Ki-1/1747R, enQuireBio™
Rabbit Monoclonal Antibody
199.00 € - 485.00 €
Specifications
Antigen | CD30 / TNFRSF8 |
---|---|
Clone | Ki-1/1747R |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15931228
|
EnquireBio
943-RBM7-P1 |
100 μg |
485.00 €
100µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15921228
|
EnquireBio
943-RBM7-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
CD30 / TNFRSF8 | |
200μg/mL | |
Monoclonal | |
Rabbit | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
943 | |
Recombinant human full-length TNFRSF8 protein | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
105-120kDa |
Ki-1/1747R | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
XP_011540745 | |
TNFRSF8 | |
IgG κ | |
Protein A/G Purified | |
Cell surface | |
Hodgkin's lymphoma |