Learn More
IBA Lifesciences CD31 Fab-Strep
Low affinity Fab fragment fused to a Twin-Strep-tag™ and specific for human CD31
Brand: IBA Lifesciences 6-8016-150
Additional Details : Weight : 0.00600kg
Description
- CD31 Fab-Strep is a recombinantly expressed low affinity Fab fragment fused to a Twin-Strep-tag™
- Due to its low affinity for the target, an increase in avidity is required to ensure sufficient binding to the cell surface
- Combining Fab-Streps with a Strep-Tactin™ backbone leads to the formation of complexes that stably bind to cells
- Fab-Streps were developed for Traceless Affinity Cell Selection (Fab-TACS™), which is based on our Strep-tag™ technology
Specifications
Lyophilized | |
Fluorescent Cell Staining & Sorting, Magnetic or Affinity Chromatographic Cell Isolation | |
Human |
For Research Use Only.