missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD32 Rat, Biotin, Clone: 2.4G2, enQuireBio™
Rat Monoclonal Antibody
94.25 € - 481.00 €
Specifications
Antigen | CD32 |
---|---|
Clone | 2.4G2 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15922588
|
EnquireBio
QAB87-B-25UG |
25 μg |
94.25 €
25µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15912588
|
EnquireBio
QAB87-B-100UG |
100 μg |
276.00 €
100µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15932588
|
EnquireBio
QAB87-B-500UG |
500 μg |
481.00 €
500µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD32 | |
0.5mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
14131 | |
IgG2b | |
Protein A/G Purified |
2.4G2 | |
Flow Cytometry | |
Biotin | |
RUO | |
XP_006496722 | |
FCGR2b | |
Primary | |
Store at 2-4°C. |